Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Eppendorf
®
CellXpert
®
C170i CO2 Incubator
Products
Product Category
heaters (52)
Manufacturer
Package Size
Filter Products
Showing 1-20 of 52
Page 1 of 3
Page 1 of 3
Compare
Product Number
Description
Pricing
EP6731000021
Eppendorf
®
CellXpert
®
C170i CO2 Incubator,
Handle on left, inner door non-segmented, AC/DC input 220—240 V, 50—60 Hz, EuroPlug
Expand
View Pricing
EP6731000351
Eppendorf
®
CellXpert
®
C170i CO2 Incubator,
water/humidity monitoring, Handle on left, inner door with 4 door segments, AC/DC input 220—240 V, 50—60 Hz, EuroPlug
Expand
View Pricing
EP6731001341
Eppendorf
®
CellXpert
®
C170i CO2 Incubator,
Oxygen control, water/humidity monitoring, Handle on right, inner door with 4 door segments, AC/DC input 220—240 V, 50—60 Hz, EuroPlug
Expand
View Pricing
EP6731001011
Eppendorf
®
CellXpert
®
C170i CO2 Incubator,
Oxygen control, Handle on right, inner door non-segmented, AC/DC input 220—240 V, 50—60 Hz, EuroPlug
Expand
View Pricing
EP6731001811
Eppendorf
®
CellXpert
®
C170i CO2 Incubator,
Oxygen control, water/humidity monitoring, Handle on right, inner door non-segmented, AC/DC input 220—240 V, 50—60 Hz, EuroPlug
Expand
View Pricing
EP6731000051
Eppendorf
®
CellXpert
®
C170i CO2 Incubator,
Handle on left, inner door with 4 door segments, AC/DC input 220—240 V, 50—60 Hz, EuroPlug
Expand
View Pricing
EP6731000011
Eppendorf
®
CellXpert
®
C170i CO2 Incubator,
Handle on right, inner door non-segmented, AC/DC input 220—240 V, 50—60 Hz, EuroPlug
Expand
View Pricing
EP6731001021
Eppendorf
®
CellXpert
®
C170i CO2 Incubator,
Oxygen control, Handle on left, inner door non-segmented, AC/DC input 220—240 V, 50—60 Hz, EuroPlug
Expand
View Pricing
EP6731001352
Eppendorf
®
CellXpert
®
C170i CO2 Incubator,
Oxygen control, water/humidity monitoring, Handle on left, inner door with 4 door segments, AC/DC input 220—240 V, 50—60 Hz, UK plug
Expand
View Pricing
EP6731001522
Eppendorf
®
CellXpert
®
C170i CO2 Incubator,
Copper interior, Oxygen control, Handle on left, inner door non-segmented, AC/DC input 220—240 V, 50—60 Hz, UK plug
Expand
View Pricing
EP6731001022
Eppendorf
®
CellXpert
®
C170i CO2 Incubator,
Oxygen control, Handle on left, inner door non-segmented, AC/DC input 220—240 V, 50—60 Hz, UK plug
Expand
View Pricing
EP6731000352
Eppendorf
®
CellXpert
®
C170i CO2 Incubator,
water/humidity monitoring, Handle on left, inner door with 4 door segments, AC/DC input 220—240 V, 50—60 Hz, UK plug
Expand
View Pricing
EP6731000311
Eppendorf
®
CellXpert
®
C170i CO2 Incubator,
water/humidity monitoring, Handle on right, inner door non-segmented, AC/DC input 220—240 V, 50—60 Hz, EuroPlug
Expand
View Pricing
EP6731000851
Eppendorf
®
CellXpert
®
C170i CO2 Incubator,
Copper interior, water/humidity monitoring, Handle on left, inner door with 4 door segments, AC/DC input 220—240 V, 50—60 Hz, EuroPlug
Expand
View Pricing
EP6731000841
Eppendorf
®
CellXpert
®
C170i CO2 Incubator,
Copper interior, water/humidity monitoring, Handle on right, inner door with 4 door segments, AC/DC input 220—240 V, 50—60 Hz, EuroPlug
Expand
View Pricing
EP6731000052
Eppendorf
®
CellXpert
®
C170i CO2 Incubator,
Handle on left, inner door with 4 door segments, AC/DC input 220—240 V, 50—60 Hz, UK plug
Expand
View Pricing
EP6731001821
Eppendorf
®
CellXpert
®
C170i CO2 Incubator,
Oxygen control, water/humidity monitoring, Handle on left, inner door non-segmented, AC/DC input 220—240 V, 50—60 Hz, EuroPlug
Expand
View Pricing
EP6731001052
Eppendorf
®
CellXpert
®
C170i CO2 Incubator,
Oxygen control, Handle on left, inner door with 4 door segments, AC/DC input 220—240 V, 50—60 Hz, UK plug
Expand
View Pricing
EP6731001091
Eppendorf
®
CellXpert
®
C170i CO2 Incubator,
Oxygen control, Handle on left, inner door with 8 door segments, AC/DC input 220—240 V, 50—60 Hz, EuroPlug
Expand
View Pricing
EP6731000121
Eppendorf
®
CellXpert
®
C170i CO2 Incubator,
humidity monitoring, Handle on left, inner door non-segmented, AC/DC input 220—240 V, 50—60 Hz, EuroPlug
Expand
View Pricing
Page 1 of 3
Page 1 of 3