Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Eppendorf
®
Buckets
Synonym(s):
Eppendorf
®
Buckets
Products
Manufacturer
Package Size
Technique
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
EP5495513000
Eppendorf
®
Buckets,
96 x 0.2-PCR (Centr 5425)
Expand
View Pricing
EP5825730003
Eppendorf
®
Buckets,
for 7 x 50 mL Falcon tubes, set of 4 pieces
Expand
View Pricing
EP5804743008
Eppendorf
®
Buckets,
2xDWP-buckets for rotor A-2-DWP, 5804/5810
Expand
View Pricing
EP5804742001
Eppendorf
®
Buckets,
2xMTP-buckets for rotor A-2-MTP, 5804/5810
Expand
View Pricing
EP5492008810
Eppendorf
®
Buckets,
fitting rotors A-12-11 & A-8-11, set of 8 pieces
Expand
View Pricing
EP5804741005
Eppendorf
®
Buckets,
for rectangular buckets 100 ml
Expand
View Pricing