Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Eppendorf
®
Bottle Thread Adapter
Synonym(s):
Eppendorf
®
Bottle Thread Adapter
Products
Product Category
adapters (15)
Manufacturer
Filter Products
Showing 1-15 of 15
Compare
Product Number
Description
Pricing
EP4960800155
Eppendorf
®
Bottle Thread Adapter,
from GL 45 to GL 38, polypropylene
Expand
View Pricing
EP4960800120
Eppendorf
®
Bottle Thread Adapter,
from GL 45 to GL 32, polypropylene
Expand
View Pricing
EP4960800147
Eppendorf
®
Bottle Thread Adapter,
from GL 45 to S 40 (buttress thread), polypropylene
Expand
View Pricing
EP4960834009
Eppendorf
®
Bottle Thread Adapter,
from GL 45 to GL 40, PTFE
Expand
View Pricing
EP4960839000
Eppendorf
®
Bottle Thread Adapter,
from GL 45 to GL 38, ETFE
Expand
View Pricing
EP4960836001
Eppendorf
®
Bottle Thread Adapter,
from GL 32 to GL 38, ETFE
Expand
View Pricing
EP4960800040
Eppendorf
®
Bottle Thread Adapter,
from GL 32 to GL 25, polypropylene
Expand
View Pricing
EP4960800074
Eppendorf
®
Bottle Thread Adapter,
from GL 32 to GL 40, polypropylene
Expand
View Pricing
EP4960800082
Eppendorf
®
Bottle Thread Adapter,
from GL 32 to NS 19/26, polypropylene
Expand
View Pricing
EP4960800058
Eppendorf
®
Bottle Thread Adapter,
from GL 32 to GL 28, polypropylene
Expand
View Pricing
EP4960800090
Eppendorf
®
Bottle Thread Adapter,
from GL 32 to NS 24/29, polypropylene
Expand
View Pricing
EP4960835005
Eppendorf
®
Bottle Thread Adapter,
from GL 32 to GL 28, ETFE
Expand
View Pricing
EP4960800139
Eppendorf
®
Bottle Thread Adapter,
from GL 32 to GL 27, polypropylene
Expand
View Pricing
EP4960800104
Eppendorf
®
Bottle Thread Adapter,
from GL 32 to NS 29/32, polypropylene
Expand
View Pricing
EP4960833002
Eppendorf
®
Bottle Thread Adapter,
from GL 32 to GL 40, ETFE
Expand
View Pricing