Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Endoproteinase Glu-C from
Staphylococcus aureus
V8
Synonym(s):
V8 Protease
CAS Number:
66676-43-5
EC Number:
200-350-6
Products
Product Category
enzymes (1)
proteases (1)
Brand
Sigma-Aldrich (2)
Physical Form
Analyte Suitability
Quality Segment
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
P2922
Endoproteinase Glu-C from
Staphylococcus aureus
V8,
Type XVII-B, lyophilized powder, 500-1,000 units/mg solid
Expand
View Pricing
P6181
Endoproteinase Glu-C from
Staphylococcus aureus
V8,
suitable for protein sequencing, lyophilized powder
Expand
View Pricing