Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Empty SPE Tube (no frits)
Synonym(s):
SPE Reservoir
Products
Product Category
tubes (6)
Brand
Supelco (6)
Volume (ml)
Quality Segment
Application
Package Size
Technique
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
57242
Empty SPE Tube (no frits),
volume 6 mL, pkg of 30 ea
Expand
View Pricing
57021
Empty SPE Tube (no frits),
volume 20 mL, pk of 12
Expand
View Pricing
57241
Empty SPE Tube (no frits),
volume 3 mL, pk of 54
Expand
View Pricing
57022
Empty SPE Tube (no frits),
volume 60 mL, pk of 12
Expand
View Pricing
57240-U
Empty SPE Tube (no frits),
volume 1 mL, pk of 108
Expand
View Pricing
57179
Empty SPE Tube (no frits),
volume 12 mL, pk of 20
Expand
View Pricing