Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Empty polypropylene SPE Tube with PE frits, 20 μm porosity
Products
Product Category
tubes (12)
Brand
Supelco (12)
Volume (ml)
Quality Segment
Application
Package Size
Pore size (µm)
Technique
Filter Products
Showing 1-12 of 12
Compare
Product Number
Description
Pricing
57023
Empty polypropylene SPE Tube with PE frits, 20 μm porosity,
volume 1 mL, pk of 108
Expand
View Pricing
57026
Empty polypropylene SPE Tube with PE frits, 20 μm porosity,
volume 6 mL, pk of 30
Expand
View Pricing
57119-U
Empty polypropylene SPE Tube with PE frits, 20 μm porosity,
volume 60 mL, pre-fritted, pk of 16
Expand
View Pricing
57118-U
Empty polypropylene SPE Tube with PE frits, 20 μm porosity,
volume 20 mL, pre-fritted, pk of 20
Expand
View Pricing
54221-U
Empty polypropylene SPE Tube with PE frits, 20 μm porosity,
volume 3 mL, pre-fritted, pk of 100
Expand
View Pricing
57178
Empty polypropylene SPE Tube with PE frits, 20 μm porosity,
volume 60 mL, pk of 16
Expand
View Pricing
54220-U
Empty polypropylene SPE Tube with PE frits, 20 μm porosity,
volume 1 mL, pre-fritted, pk of 100
Expand
View Pricing
54222-U
Empty polypropylene SPE Tube with PE frits, 20 μm porosity,
volume 6 mL, pre-fritted, pk of 100
Expand
View Pricing
57177
Empty polypropylene SPE Tube with PE frits, 20 μm porosity,
volume 20 mL, pk of 20
Expand
View Pricing
54223-U
Empty polypropylene SPE Tube with PE frits, 20 μm porosity,
volume 12 mL, pre-fritted, pk of 100
Expand
View Pricing
57024
Empty polypropylene SPE Tube with PE frits, 20 μm porosity,
volume 3 mL, pk of 54
Expand
View Pricing
57176
Empty polypropylene SPE Tube with PE frits, 20 μm porosity,
volume 12 mL, pk of 20
Expand
View Pricing