Skip to Content
Merck
D-Panose analytical standard

D-Panose

D-Panose analytical standard
Synonym(s):
α-D-Glc-(1→6)-α-D-Glc-(1→4)-D-Glc, O-α-D-Glucopyranosyl-(1→6)-O-α-D-glucopyranosyl-(1→4)-D-glucose
Empirical Formula (Hill Notation):
C18H32O16
CAS Number:
Molecular Weight:
504.44
EC Number:

Products

Product Category
Formula Weight
Purity
Format
Quality Segment
Application
Optical Isomer
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing