Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Dodecyltrimethylammonium bromide
Synonym(s):
Lauryltrimethylammonium bromide
Linear Formula:
CH
3
(CH
2
)
11
N(CH
3
)
3
Br
CAS Number:
1119-94-4
Molecular Weight:
308.34
EC Number:
214-290-3
Products
Product Category
detergents (3)
Biological Source
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Technique
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
D8638
Dodecyltrimethylammonium bromide,
≥98%
Expand
View Pricing
D5047
Dodecyltrimethylammonium bromide,
BioXtra, ~99%
Expand
View Pricing
44239
Dodecyltrimethylammonium bromide,
suitable for ion pair chromatography, LiChropur
™
, ≥98.5% (AT)
Expand
View Pricing