Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
(±)-Geosmin solution
Empirical Formula (Hill Notation):
C
12
H
22
O
CAS Number:
16423-19-1
Molecular Weight:
182.30
EC Number:
200-659-6
Products
Product Category
building blocks (1)
Brand
Sigma-Aldrich (1)
Supelco (2)
Formula Weight
Product Line
Purity
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
certified reference material (2)
Technique
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
CRM47522
(±)-Geosmin solution,
certified reference material,
Trace
CERT
®
, 100 μg/mL in methanol, ampule of 1 mL
Expand
View Pricing
G5908
(±)-Geosmin solution,
2 mg/mL in methanol, ≥97%
Expand
View Pricing
4M7522-U
(±)-Geosmin solution,
certified reference material, 100 μg/mL in methanol
Expand
View Pricing