Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Disposable polystyrene weighing dishes
Products
Product Category
dishes (6)
Brand
Sigma-Aldrich (1)
Color
Manufacturer
Height (mm)
Length (mm)
Package Size
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
Z154873
Disposable polystyrene weighing dishes,
L × W × H 40 mm × 40 mm × 8 mm, white
Expand
View Pricing
Z154881
Disposable polystyrene weighing dishes,
L × W × H 86 mm × 86 mm × 25 mm, white
Expand
View Pricing
Z154903
Disposable polystyrene weighing dishes,
white, L × W × H 135 mm × 135 mm × 20 mm
Expand
View Pricing
Z186856
Disposable polystyrene weighing dishes,
blue, L × W × H 40 mm × 40 mm × 8 mm
Expand
View Pricing
Z186864
Disposable polystyrene weighing dishes,
L × W × H 86 mm × 86 mm × 25 mm, blue
Expand
View Pricing
Z186872
Disposable polystyrene weighing dishes,
blue, L × W × H 135 mm × 135 mm × 20 mm
Expand
View Pricing