
(+)-Isopulegol

Synonym(s):
(1S,2R,5S)-2-Isopropenyl-5-methylcyclohexanol, (1S,3S,4R)-p-Menth-8-en-3-ol
Empirical Formula (Hill Notation):
C10H18O
CAS Number:
Molecular Weight:
154.25
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Format
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing