Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Dimethylpolysiloxane
Synonym(s):
Polydimethylsiloxane
CAS Number:
9016-00-6
Products
Brand
Sigma-Aldrich (4)
Supelco (1)
Physical Form
Format
Quality Segment
Special Grade
analytical standard (1)
Technique
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
DMPS5C
Dimethylpolysiloxane,
viscosity 500 cSt (25 °C)(lit.)
Expand
View Pricing
DMPSV
Dimethylpolysiloxane,
viscosity 5 cSt (25 °C)(lit.)
Expand
View Pricing
DMPS2C
Dimethylpolysiloxane,
viscosity 200 cSt (25 °C)(lit.)
Expand
View Pricing
DMPS5X
Dimethylpolysiloxane,
viscosity 50 cSt (25 °C)(lit.)
Expand
View Pricing
361879
Dimethylpolysiloxane,
set of analytical standards, molecular weight series
Expand
View Pricing