
Dimethyl aminomalonate hydrochloride

Synonym(s):
Aminomalonic acid dimethyl ester hydrochloride
Linear Formula:
NH2CH(COOCH3)2 · HCl
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing