
Dihydrotachysterol for system suitability

Synonym(s):
Dihydrotachysterol, 9,10-Secoergosta-5,7,22-trien-3β-ol, DHT2
Empirical Formula (Hill Notation):
C28H46O
CAS Number:
Molecular Weight:
398.66
Products
Product Category
Biological Source
Formula Weight
Manufacturer
Physical Form
Agency
Format
Application
API Family
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing