Skip to Content
Merck
Diethylene glycol diethyl ether suitable for HPLC, ≥99%

Diethylene glycol diethyl ether

Diethylene glycol diethyl ether suitable for HPLC, ≥99%
Synonym(s):
2-Ethoxyethyl ether, Bis(2-ethoxyethyl) ether, Diethyldiglycol, Diethylene glycol diethyl ether
Linear Formula:
(C2H5OCH2CH2)2O
CAS Number:
Molecular Weight:
162.23

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Application
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing