
Didanosine Related Compound A

Synonym(s):
Hypoxanthine, 6-Hydroxypurine
Empirical Formula (Hill Notation):
C5H4N4O
CAS Number:
Molecular Weight:
136.11
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing