Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Dialysis tubing closures
Products
Brand
Sigma-Aldrich (15)
Color
Filter Products
Showing 1-17 of 17
Compare
Product Number
Description
Pricing
Z371092
Dialysis tubing closures,
blue, size 50 mm
Expand
View Pricing
Z371106
Dialysis tubing closures,
blue, size 70 mm
Expand
View Pricing
Z370991
Dialysis tubing closures,
red, size 50 mm
Expand
View Pricing
Z370959
Dialysis tubing closures,
white, size 50 mm
Expand
View Pricing
Z371041
Dialysis tubing closures,
yellow, size 50 mm
Expand
View Pricing
Z371130
Dialysis tubing closures,
green, size 50 mm
Expand
View Pricing
Z371165
Dialysis tubing closures,
black, size 70 mm
Expand
View Pricing
Z371157
Dialysis tubing closures,
black, size 50 mm
Expand
View Pricing
Z371025
Dialysis tubing closures,
red, size 150 mm
Expand
View Pricing
Z371009
Dialysis tubing closures,
red, size 70 mm
Expand
View Pricing
Z371068
Dialysis tubing closures,
yellow, size 70 mm
Expand
View Pricing
Z370967
Dialysis tubing closures,
white, size 70 mm
Expand
View Pricing
Z370975
Dialysis tubing closures,
white, size 110 mm
Expand
View Pricing
Z371017
Dialysis tubing closures,
red, size 110 mm
Expand
View Pricing
Z371084
Dialysis tubing closures,
yellow, size 150 mm
Expand
View Pricing
Z371076
Dialysis tubing closures,
yellow, size 110 mm
Expand
View Pricing
Z371149
Dialysis tubing closures,
green, size 70 mm
Expand
View Pricing