
D-Glucose-1-13C

Synonym(s):
Dextrose-1-13C
Empirical Formula (Hill Notation):
13CC5H12O6
CAS Number:
Molecular Weight:
181.15
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Isotope
Markush Class
Markush Group
Optical Isomer
Special Grade
Technique
Showing 1-3 of 3