
Dextromethorphan Impurity C

Synonym(s):
(9α,13α,14α)-3-Methoxy-17-methylmorphinan-10-one, 10-Keto dextromethorphan
Empirical Formula (Hill Notation):
C18H23NO2
CAS Number:
Molecular Weight:
285.38
Products
Brand
Formula Weight
Agency
Format
Quality Segment
Application
Package Size
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing