Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Dextran from
Leuconostoc mesenteroides
Linear Formula:
(C
6
H
10
O
5
)
n
CAS Number:
9004-54-0
EC Number:
232-677-5
Products
Product Category
carbohydrates (9)
dextrans (7)
polysaccharides (7)
Brand
Sigma-Aldrich (9)
Supelco (6)
Biological Source
Color
Physical Form
Format
Analyte Suitability
Research Area
Quality Segment
Application
Reference Material Type
Special Grade
analytical standard (7)
Technique
Filter Products
Showing 1-16 of 16
Compare
Product Number
Description
Pricing
D9260
Dextran from
Leuconostoc mesenteroides
,
average mol wt 9,000-11,000
Expand
View Pricing
D5376
Dextran from
Leuconostoc mesenteroides
,
average mol wt 1,500,000-2,800,000
Expand
View Pricing
D8821
Dextran from
Leuconostoc mesenteroides
,
average mol wt 60,000-76,000
Expand
View Pricing
D1662
Dextran from
Leuconostoc mesenteroides
,
average mol wt 35,000-45,000
Expand
View Pricing
D4876
Dextran from
Leuconostoc mesenteroides
,
average mol wt 150,000
Expand
View Pricing
31398
Dextran from
Leuconostoc mesenteroides
,
M
r
~200,000
Expand
View Pricing
31397
Dextran from
Leuconostoc mesenteroides
,
M
r
~60,000
Expand
View Pricing
31425
Dextran from
Leuconostoc mesenteroides
,
analytical standard, for GPC, Mw 670,000
Expand
View Pricing
D3759
Dextran from
Leuconostoc mesenteroides
,
average mol wt 48,000-90,000
Expand
View Pricing
31417
Dextran from
Leuconostoc mesenteroides
,
analytical standard, for GPC, Mw 5,000
Expand
View Pricing
D4626
Dextran from
Leuconostoc mesenteroides
,
average mol wt 15,000-30,000
Expand
View Pricing
31416
Dextran from
Leuconostoc mesenteroides
,
analytical standard, for GPC, Mw 1,000
Expand
View Pricing
31419
Dextran from
Leuconostoc mesenteroides
,
analytical standard, for GPC, Mw 25,000
Expand
View Pricing
31420
Dextran from
Leuconostoc mesenteroides
,
for GPC, analytical standard, Mw 50,000
Expand
View Pricing
31424
Dextran from
Leuconostoc mesenteroides
,
analytical standard, for GPC, Mw 410,000
Expand
View Pricing
31418
Dextran from
Leuconostoc mesenteroides
,
analytical standard, for GPC, Mw 12,000
Expand
View Pricing