![D-Erythrose 4-phosphate sodium salt ≥50% (TLC)](/deepweb/assets/sigmaaldrich/product/structures/383/147/9e2e27ab-fa79-4723-86d9-7bee5b27f155/640/9e2e27ab-fa79-4723-86d9-7bee5b27f155.png)
D-Erythrose 4-phosphate sodium salt
![D-Erythrose 4-phosphate sodium salt ≥50% (TLC)](/deepweb/assets/sigmaaldrich/product/structures/383/147/9e2e27ab-fa79-4723-86d9-7bee5b27f155/640/9e2e27ab-fa79-4723-86d9-7bee5b27f155.png)
Synonym(s):
4-Phospho-D-erythrose sodium salt
Empirical Formula (Hill Notation):
C4H8NaO7P
CAS Number:
Molecular Weight:
222.07
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Quality Segment
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing