Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Deoxyribonuclease I from bovine pancreas
Synonym(s):
DNase I, Deoxyribonucleate 5′-oligonucleotido-hydrolase
CAS Number:
9003-98-9
EC Number:
232-667-0
Products
Product Category
enzymes (5)
Brand
Sigma-Aldrich (5)
Biological Source
Physical Form
Tissue
Chemical Composition
Application
Package Size
Sterilization Method
Technique
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
DN25
Deoxyribonuclease I from bovine pancreas,
lyophilized powder, Protein ≥85 %, ≥400 Kunitz units/mg protein
Expand
View Pricing
D5025
Deoxyribonuclease I from bovine pancreas,
Type IV, lyophilized powder, ≥2,000 Kunitz units/mg protein
Expand
View Pricing
D4527
Deoxyribonuclease I from bovine pancreas,
Type II, lyophilized powder, Protein ≥80 %, ≥2,000 units/mg protein
Expand
View Pricing
D4513
Deoxyribonuclease I from bovine pancreas,
Type II-S, lyophilized powder, Protein ≥80 %, ≥2,000 units/mg protein
Expand
View Pricing
D4263
Deoxyribonuclease I from bovine pancreas,
Standardized vial containing 2,000 Kunitz units of DNase I (D4527), vial of ≥0.25 mg total protein
Expand
View Pricing