Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Density Standard (20 °C)
Products
Manufacturer
Agency
Format
Quality Segment
Application
Reference Material Type
Special Grade
certified reference material (12)
Filter Products
Showing 1-12 of 12
Compare
Product Number
Description
Pricing
DEN2011
Density Standard (20 °C),
UKAS ISO/IEC17025 and ISO 17034 certified, density: 1.1258 g/mL at 20 °C
Expand
View Pricing
DEN2003
Density Standard (20 °C),
UKAS ISO/IEC17025 and ISO 17034 certified, density: 0.7788 g/mL at 20 °C
Expand
View Pricing
DEN2013
Density Standard (20 °C),
UKAS ISO/IEC17025 and ISO 17034 certified, density: 1.2849 g/mL at 20 °C
Expand
View Pricing
DEN2012
Density Standard (20 °C),
UKAS ISO/IEC17025 and ISO 17034 certified, density: 1.1917 g/mL at 20 °C
Expand
View Pricing
DEN2014
Density Standard (20 °C),
UKAS ISO/IEC17025 and ISO 17034 certified, density: 1.6226 g/mL at 20 °C
Expand
View Pricing
DEN2004
Density Standard (20 °C),
UKAS ISO/IEC17025 and ISO 17034 certified, density: 0.8383 g/mL at 20 °C
Expand
View Pricing
DEN2002
Density Standard (20 °C),
UKAS ISO/IEC17025 and ISO 17034 certified, density: 0.7264 g/mL at 20 °C
Expand
View Pricing
DEN2007
Density Standard (20 °C),
UKAS ISO/IEC17025 and ISO 17034 certified, density: 0.9376 g/mL at 20 °C
Expand
View Pricing
DEN2010
Density Standard (20 °C),
UKAS ISO/IEC17025 and ISO 17034 certified, density: 1.0677 g/mL at 20 °C
Expand
View Pricing
DEN2006
Density Standard (20 °C),
UKAS ISO/IEC17025 and ISO 17034 certified, density: 0.8708 g/mL at 20 °C
Expand
View Pricing
DEN2008
Density Standard (20 °C),
UKAS ISO/IEC17025 and ISO 17034 certified, density: 0.9817 g/mL at 20 °C
Expand
View Pricing
DEN2009
Density Standard (20 °C),
UKAS ISO/IEC17025 and ISO 17034 certified, density: 1.0257 g/mL at 20 °C
Expand
View Pricing