Skip to Content
Merck
Delta-9-Tetrahydrocannabinol United States Pharmacopeia (USP) Reference Standard

Delta-9-Tetrahydrocannabinol

Delta-9-Tetrahydrocannabinol United States Pharmacopeia (USP) Reference Standard
Synonym(s):
Δ9-Tetrahydrocannabinol solution, Δ1-Tetrahydrocannabinol
Empirical Formula (Hill Notation):
C21H30O2
CAS Number:
Molecular Weight:
314.46

Products

Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1