Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
DCI Activator
Synonym(s):
DCI activator solution
Products
Product Category
nucleosides (2)
Brand
Sigma-Aldrich (4)
Color
Physical Form
Product Line
Quality Segment
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
L032000
DCI Activator,
0.25 M
Expand
View Pricing
L032100
DCI Activator,
0.5 M
Expand
View Pricing
L032080
DCI Activator,
0.25 M, configured for PerkinElmer, configured for Polygen
Expand
View Pricing
L032030
DCI Activator,
0.25 M, configured for ABI
Expand
View Pricing