Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
D-α-Tocopherol succinate
Synonym(s):
Vitamin E succinate
Empirical Formula (Hill Notation):
C
33
H
54
O
5
CAS Number:
4345-03-3
Molecular Weight:
530.78
EC Number:
224-403-8
Products
Product Category
vitamins (3)
Brand
Sigma-Aldrich (2)
Supelco (1)
Biological Source
Color
Formula Weight
Physical Form
Product Line
Purity
Format
Application
Optical Isomer
Package Size
Reference Material Type
Special Grade
analytical standard (1)
Technique
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
T3126
D-α-Tocopherol succinate,
semisynthetic, 1210 IU/g
Expand
View Pricing
47782
D-α-Tocopherol succinate,
analytical standard
Expand
View Pricing
95255
D-α-Tocopherol succinate,
BioXtra, ≥98.0% (HPLC)
Expand
View Pricing