Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Cytochrome c from equine heart
Synonym(s):
Cytochrome c from horse heart
CAS Number:
9007-43-6
EC Number:
232-700-9
Products
Product Category
proteins (3)
Brand
Sigma-Aldrich (4)
Biological Source
Physical Form
Product Line
Purity
Tissue
Quality Segment
Application
Package Size
Technique
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
C2506
Cytochrome c from equine heart,
≥95% (SDS-PAGE)
Expand
View Pricing
C7752
Cytochrome c from equine heart,
≥95% based on Mol. Wt. 12,384 basis
Expand
View Pricing
C7150
Cytochrome c from equine heart,
suitable for GFC marker, BioReagent
Expand
View Pricing
C2867
Cytochrome c from equine heart,
BioUltra, ≥99% (SDS-PAGE), powder, suitable for mammalian cell culture
Expand
View Pricing