Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Cymarin
Synonym(s):
K-Strophanthidin-D-cymaroside, K-Strophanthin-α
Empirical Formula (Hill Notation):
C
30
H
44
O
9
CAS Number:
508-77-0
Molecular Weight:
548.66
EC Number:
208-087-9
Products
Product Category
lipids (1)
metabolites (1)
steroids (1)
Brand
Sigma-Aldrich (1)
Formula Weight
Manufacturer
Product Line
Purity
Format
Quality Segment
Application
Optical Isomer
Reference Material Type
Special Grade
analytical standard (1)
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
30030
Cymarin,
≥96% (HPLC)
Expand
View Pricing
PHL89838
Cymarin,
phyproof
®
Reference Substance
Expand
View Pricing