![Cyclopropylboronic acid](/deepweb/assets/sigmaaldrich/product/structures/275/825/bb2385b5-a125-42d1-814c-50d637f6d7f6/640/bb2385b5-a125-42d1-814c-50d637f6d7f6.png)
Cyclopropylboronic acid
![Cyclopropylboronic acid](/deepweb/assets/sigmaaldrich/product/structures/275/825/bb2385b5-a125-42d1-814c-50d637f6d7f6/640/bb2385b5-a125-42d1-814c-50d637f6d7f6.png)
Synonym(s):
Cyclopropaneboronic acid
Empirical Formula (Hill Notation):
C3H7BO2
CAS Number:
Molecular Weight:
85.90
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing