Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Cyclohexene
Synonym(s):
Cyclohexene, Tetrahydrobenzene
Empirical Formula (Hill Notation):
C
6
H
10
CAS Number:
110-83-8
Molecular Weight:
82.14
EC Number:
203-807-8
Products
Product Category
building blocks (1)
solvents (1)
Brand
Sigma-Aldrich (3)
Supelco (1)
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Purum (1)
analytical standard (1)
Technique
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
8.02824
Cyclohexene,
(stabilized) for synthesis
Expand
View Pricing
125431
Cyclohexene,
inhibitor-free,
ReagentPlus
®
, 99%
Expand
View Pricing
44028
Cyclohexene,
analytical standard, ≥99.5% (GC)
Expand
View Pricing
29240
Cyclohexene,
contains 100 ppm BHT as inhibitor, ≥99.0%
Expand
View Pricing