
Cyanidin chloride

Synonym(s):
3,3′,4,5,7-Pentahydroxyflavylium chloride, Cyanidol chloride
Empirical Formula (Hill Notation):
C15H11ClO6
CAS Number:
Molecular Weight:
322.70
Products
Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-6 of 6
Compare
Product Number
Description
Pricing