Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
CryoFile
®
XL freezer box, for batching and storing cryogenic vials or ampules
Products
Product Category
boxes (6)
Color
Height (mm)
Length (mm)
Technique
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
Z764957
CryoFile
®
XL freezer box, for batching and storing cryogenic vials or ampules,
white
Expand
View Pricing
Z764965
CryoFile
®
XL freezer box, for batching and storing cryogenic vials or ampules,
blue
Expand
View Pricing
Z764949
CryoFile
®
XL freezer box, for batching and storing cryogenic vials or ampules,
pink
Expand
View Pricing
Z764973
CryoFile
®
XL freezer box, for batching and storing cryogenic vials or ampules,
red
Expand
View Pricing
Z764922
CryoFile
®
XL freezer box, for batching and storing cryogenic vials or ampules,
green
Expand
View Pricing
Z764930
CryoFile
®
XL freezer box, for batching and storing cryogenic vials or ampules,
yellow
Expand
View Pricing