Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Crimp seals (magnetic) with PTFE/silicone septa, pkg/100
Products
Product Category
septa (5)
Brand
Supelco (4)
Diameter (mm)
Compatibility
Closure Type
Color
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
SU860015
Crimp seals (magnetic) with PTFE/silicone septa, pkg/100,
gold seal (magnetic, with 8 mm center hole), (silcone blue transparent/PTFE transparent), open center (8 mm opening), diam. × thickness 20 mm × 3.0 mm
Expand
View Pricing
SU860053
Crimp seals (magnetic) with PTFE/silicone septa, pkg/100,
gold seal (magnetic, with 8 mm center hole), PTFE/silicone (blue PTFE/white silicone), diam. × thickness 20 mm × 1.5 mm, open center
Expand
View Pricing
27300
Crimp seals (magnetic) with PTFE/silicone septa, pkg/100,
silver steel (magnetic, with 5 mm center hole), PTFE/silicone, seal diam. × total thickness 20 mm × 3 mm
Expand
View Pricing
SU860105
Crimp seals (magnetic) with PTFE/silicone septa, pkg/100,
gold seal (magnetic, with 8 mm center hole), (Silicone blue transparent/PTFE tranparent), diam. × thickness 20 mm × 3.0 mm, open center
Expand
View Pricing
854179-U
Crimp seals (magnetic) with PTFE/silicone septa, pkg/100,
gold seal (magnetic, with 5 mm center hole), (silcone blue transparent/PTFE transparent), diam. × thickness 20 mm × 3.0 mm, open center
Expand
View Pricing