Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Corning
®
syringe filters
Products
Product Category
syringe filters (11)
Manufacturer
Diameter (mm)
Housing Material
Membrane Material
Package Size
Pore size (µm)
Sterility
Filter Products
Showing 1-11 of 11
Compare
Product Number
Description
Pricing
CLS431219
Corning
®
syringe filters,
cellulose acetate membrane (surfactant-free), diam. 28 mm, pore size 0.2 μm
Expand
View Pricing
CLS431229
Corning
®
syringe filters,
polyethersulfone membrane, pore size 0.2 μm
Expand
View Pricing
CLS431220
Corning
®
syringe filters,
cellulose acetate membrane (surfactant-free), diam. 28 mm, pore size 0.45 μm
Expand
View Pricing
CLS431224
Corning
®
syringe filters,
Nylon membrane, diam. 25 mm, pore size 0.2 μm
Expand
View Pricing
CLS431215
Corning
®
syringe filters,
regenerated cellulose membrane, diam. 15 mm, pore size 0.2 μm
Expand
View Pricing
CLS431225
Corning
®
syringe filters,
Nylon membrane, pore size 0.45 μm
Expand
View Pricing
CLS431231
Corning
®
syringe filters,
PTFE membrane, diam. 25 mm, pore size 0.45 μm
Expand
View Pricing
CLS431227
Corning
®
syringe filters,
PTFE membrane, diam. 50 mm, pore size 0.2 μm
Expand
View Pricing
CLS431218
Corning
®
syringe filters,
sterile, cellulose membrane (surfactant-free with prefilter), diam. 28 mm, pore size 0.2 μm
Expand
View Pricing
CLS431222
Corning
®
syringe filters,
regenerated cellulose membrane, diam. 25 mm, pore size 0.2 μm
Expand
View Pricing
CLS431221
Corning
®
syringe filters,
cellulose acetate membrane (surfactant-free), diam. 28 mm, pore size 0.8 μm
Expand
View Pricing