Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Corning
®
Stripettor
™
Ultra Pipet Controller Accessories
Products
Product Category
racks (1)
Manufacturer
Color
Package Size
Pore size (µm)
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
CLS4991
Corning
®
Stripettor
™
Ultra Pipet Controller Accessories,
pore size 0.2 μm, pack of 5
Expand
View Pricing
CLS4992
Corning
®
Stripettor
™
Ultra Pipet Controller Accessories,
PTFE filter (hydrophobic), pore size 0.45 μm, pack of 5
Expand
View Pricing
CLS4994
Corning
®
Stripettor
™
Ultra Pipet Controller Accessories,
Battery Set
Expand
View Pricing
CLS4996
Corning
®
Stripettor
™
Ultra Pipet Controller Accessories,
Power Supply, AC/DC input 120 - 240 V AC, universal plug set (Europlug, UK, US, and Australian Plugs)
Expand
View Pricing
CLS4993
Corning
®
Stripettor
™
Ultra Pipet Controller Accessories,
Silicone Pipet Holder
Expand
View Pricing
CLS4995
Corning
®
Stripettor
™
Ultra Pipet Controller Accessories,
Battery Cover
Expand
View Pricing
CLS4997
Corning
®
Stripettor
™
Ultra Pipet Controller Accessories,
Colored Nose Pieces, pack of 4
Expand
View Pricing