Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Corning
®
square PET storage bottles
Products
Product Category
bottles (4)
Manufacturer
Capacity (mL)
Diameter (mm)
Cap
Bottle material
Color
Vial Bottle Material
Package Size
Sterility
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
CLS431531
Corning
®
square PET storage bottles,
square clear PET, sterile, capacity 250 mL, screw cap, case of 24
Expand
View Pricing
CLS431532
Corning
®
square PET storage bottles,
square clear PET, sterile, capacity 500 mL, screw cap, case of 24
Expand
View Pricing
CLS431533
Corning
®
square PET storage bottles,
square clear PET, sterile, capacity 1000 mL, screw cap, case of 24
Expand
View Pricing
CLS431530
Corning
®
square PET storage bottles,
square clear PET, sterile, capacity 125 mL, screw cap, case of 24
Expand
View Pricing