Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Corning
®
multiwell plates, plate lids and sealing mats
Products
Product Category
plates (6)
Color
Manufacturer
Wells
Plate Material
Package Size
Sterility
Technique
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
CLS3099
Corning
®
multiwell plates, plate lids and sealing mats,
universal multiwell plate lid w/ corner notch, polystyrene, clear, sterile, 50/cs
Expand
View Pricing
CLS3931
Corning
®
multiwell plates, plate lids and sealing mats,
96 well plate lid, rigid polystyrene low evaporation lid w/ corner notch, condensation rings, sterile, 25/cs
Expand
View Pricing
CLS3098
Corning
®
multiwell plates, plate lids and sealing mats,
universal multiwell plate lid w/o corner notch, polystyrene, clear, sterile, 100/cs
Expand
View Pricing
CLS3930
Corning
®
multiwell plates, plate lids and sealing mats,
96 well plate lid, rigid polystyrene low evaporation lid w/ corner notch, condensation rings, sterile, 100/cs
Expand
View Pricing
CLS3935
Corning
®
multiwell plates, plate lids and sealing mats,
universal multiwell plate lid w/ corner notch, polystyrene, black, sterile, 50/cs
Expand
View Pricing
CLS6520
Corning
®
multiwell plates, plate lids and sealing mats,
96 well polycarbonate PCR plate lid, non-sterile, 25/cs
Expand
View Pricing