Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Corning
®
microplate sealing tape
Products
Product Category
sealing films and foils (5)
Color
Manufacturer
Length (cm)
Length (in)
Wells
Application
Color
Package Size
Sterility
Technique
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
CLS6570
Corning
®
microplate sealing tape,
aluminum film, non-sterile, suitable for (for short and long term storage and PCR, 96 well reactions)
Expand
View Pricing
CLS3345
Corning
®
microplate sealing tape,
white Rayon film (with acrylic), sterile, suitable for (for cell/tissue culture applications, breathable sterile membrane)
Expand
View Pricing
CLS6524
Corning
®
microplate sealing tape,
polyethylene film, non-sterile, suitable for (for short and long term storage and PCR, 96 well reactions)
Expand
View Pricing
CLS4612
Corning
®
microplate sealing tape,
ELISA/General Incubation, polyester film, non-sterile
Expand
View Pricing
CLS6569
Corning
®
microplate sealing tape,
aluminum film, non-sterile, pack of 100
Expand
View Pricing