Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Corning
®
gel-loading pipet tips
Products
Product Category
pipette tips (4)
Manufacturer
Length (mm)
Color
Package Size
Special Grade
Natural (3)
Sterility
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
CLS4853
Corning
®
gel-loading pipet tips,
1-200μL round 0.5mm thick gel-loading pipet tips, natural, non-sterile, 200 tips/rack, 2 racks/case, 400 tips/case
Expand
View Pricing
CLS4815
Corning
®
gel-loading pipet tips,
0.2-10μL flat microvolume gel-loading pipet tips, natural, non-sterile, 200 tips/rack, 400 tips/case
Expand
View Pricing
CLS4884
Corning
®
gel-loading pipet tips,
1 - 200 μL, end of tip thickness 0.2 mm, tip flat
Expand
View Pricing
CLS4854
Corning
®
gel-loading pipet tips,
1-200μL flat 0.4mm thick gel-loading pipet tips, natural, non-sterile, 200 tips/rack, 2 racks/case, 400 tips/case
Expand
View Pricing