Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Corning
®
Falcon
®
Permeable Support
Synonym(s):
Permeable Support
Products
Product Category
plates (18)
Manufacturer
Package Size
Sterility
Filter Products
Showing 1-18 of 18
Compare
Product Number
Description
Pricing
CLS353097
Corning
®
Falcon
®
Permeable Support,
for use with 24 Well Plate, with 8.0 µm Transparent PET Membrane, sterile
Expand
View Pricing
CLS353095
Corning
®
Falcon
®
Permeable Support,
for use with 24 Well Plate, with 0.4 µm Transparent PET Membrane, sterile
Expand
View Pricing
CLS353102
Corning
®
Falcon
®
Permeable Support,
for use with 6 Well Plate, with 1.0µm Transparent PET Membrane, sterile
Expand
View Pricing
CLS353104
Corning
®
Falcon
®
Permeable Support,
for use with 24 Well Plate, with 1.0 µm Transparent PET Membrane, sterile
Expand
View Pricing
CLS353090
Corning
®
Falcon
®
Permeable Support,
for use with 6 Well Plate, with 0.4 µm Pore Transparent PET Membrane, sterile
Expand
View Pricing
CLS353180
Corning
®
Falcon
®
Permeable Support,
for use with 12 Well Plate, with 0.4 µm Transparent PET Membrane, sterile
Expand
View Pricing
CLS353091
Corning
®
Falcon
®
Permeable Support,
for use with 6 Well Plate, with 3.0µm Translucent PET Membrane, sterile
Expand
View Pricing
CLS353494
Corning
®
Falcon
®
Permeable Support,
for use with 12 Well Plate, with 0.4 µm Translucent PET Membrane, sterile
Expand
View Pricing
CLS353096
Corning
®
Falcon
®
Permeable Support,
for use with 24 Well Plate, with 3 µm Transparent PET Membrane, sterile
Expand
View Pricing
CLS353492
Corning
®
Falcon
®
Permeable Support,
for use with 24 Well Plate, with 3 µm Translucent PET Membrane, sterile
Expand
View Pricing
CLS353182
Corning
®
Falcon
®
Permeable Support,
for use with 12 Well Plate, with 8.0 µm Transparent PET Membrane, sterile
Expand
View Pricing
CLS353103
Corning
®
Falcon
®
Permeable Support,
for use with 12 Well Plate, with 1.0µm Transparent PET Membrane, sterile
Expand
View Pricing
CLS353181
Corning
®
Falcon
®
Permeable Support,
for use with 12 Well Plate, with 3 µm Transparent PET Membrane, sterile
Expand
View Pricing
CLS353495
Corning
®
Falcon
®
Permeable Support,
Expand
View Pricing
CLS353092
Corning
®
Falcon
®
Permeable Support,
for use with 6 Well Plate, with 3.0µm Translucent PET Membrane, sterile
Expand
View Pricing
CLS353093
Corning
®
Falcon
®
Permeable Support,
for use with 6 Well Plate, with 8.0µm Transparent PET Membrane, sterile
Expand
View Pricing
CLS353292
Corning
®
Falcon
®
Permeable Support,
for use with 12 Well Plate, with 3.0µm Translucent PET Membrane, sterile
Expand
View Pricing
CLS353493
Corning
®
Falcon
®
Permeable Support,
for use with 6 Well Plate, with 0.4 µm Translucent PET Membrane, sterile
Expand
View Pricing