Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Corning
®
Falcon
®
Cell Culture Plate
Synonym(s):
Cell Culture Plate
Products
Product Category
plates (16)
Manufacturer
Binding Type
Color
Package Size
Sterility
Filter Products
Showing 1-16 of 16
Compare
Product Number
Description
Pricing
CLS353046
Corning
®
Falcon
®
Cell Culture Plate,
wells, 6, Tissue Culture (TC)-treated surface, flat bottom clear wells, sterile
Expand
View Pricing
CLS353072
Corning
®
Falcon
®
Cell Culture Plate,
wells, 96, Tissue Culture (TC)-treated surface, flat bottom clear wells, sterile
Expand
View Pricing
CLS353047
Corning
®
Falcon
®
Cell Culture Plate,
wells, 24, Tissue Culture (TC)-treated surface, flat bottom clear wells, sterile
Expand
View Pricing
CLS353043
Corning
®
Falcon
®
Cell Culture Plate,
wells, 12, Tissue Culture (TC)-treated surface, flat bottom clear wells, sterile
Expand
View Pricing
CLS353077
Corning
®
Falcon
®
Cell Culture Plate,
wells, 96, Tissue Culture (TC)-treated surface, round bottom clear wells, sterile
Expand
View Pricing
CLS353078
Corning
®
Falcon
®
Cell Culture Plate,
wells, 48, Tissue Culture (TC)-treated surface, flat bottom clear wells, sterile
Expand
View Pricing
CLS353075
Corning
®
Falcon
®
Cell Culture Plate,
wells, 96, Tissue Culture (TC)-treated surface, flat bottom clear wells, sterile
Expand
View Pricing
CLS353226
Corning
®
Falcon
®
Cell Culture Plate,
wells, 24, Tissue Culture (TC)-treated surface, flat bottom clear wells, sterile
Expand
View Pricing
CLS353224
Corning
®
Falcon
®
Cell Culture Plate,
wells, 6, Tissue Culture (TC)-treated surface, flat bottom clear wells, sterile
Expand
View Pricing
CLS353227
Corning
®
Falcon
®
Cell Culture Plate,
wells, 96, Tissue Culture (TC)-treated surface, round bottom clear wells, sterile
Expand
View Pricing
CLS353934
Corning
®
Falcon
®
Cell Culture Plate,
wells, 6, Tissue Culture (TC)-treated surface, flat bottom clear wells, sterile
Expand
View Pricing
CLS353225
Corning
®
Falcon
®
Cell Culture Plate,
wells, 12, Tissue Culture (TC)-treated surface, flat bottom clear wells, sterile
Expand
View Pricing
CLS353935
Corning
®
Falcon
®
Cell Culture Plate,
wells, 24, Tissue Culture (TC)-treated surface, flat bottom clear wells, sterile
Expand
View Pricing
CLS353230
Corning
®
Falcon
®
Cell Culture Plate,
wells, 48, Tissue Culture (TC)-treated surface, flat bottom clear wells, sterile
Expand
View Pricing
CLS353936
Corning
®
Falcon
®
Cell Culture Plate,
wells, 96, Tissue Culture (TC)-treated surface, flat bottom clear wells, sterile
Expand
View Pricing
CLS353916
Corning
®
Falcon
®
Cell Culture Plate,
wells, 96, Tissue Culture (TC)-treated surface, flat bottom clear wells, sterile
Expand
View Pricing