Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Corning
®
DeckWorks
™
standard pipet tips
Products
Product Category
pipette tips (19)
Manufacturer
Binding Type
Volume Range (µL)
Color
Package Size
Special Grade
Natural (15)
Sterility
Filter Products
Showing 1-19 of 19
Compare
Product Number
Description
Pricing
CLS4114
Corning
®
DeckWorks
™
standard pipet tips,
50 - 1250μL extended length pipet tips, graduated, natural, non-sterile, polypropylene, 768 tips/bag, 7680 tips/case
Expand
View Pricing
CLS4126
Corning
®
DeckWorks
™
standard pipet tips,
1 - 200μL pipet tips, graduated, reload system, natural, non-sterile, polypropylene, 96 tips/deck, 10 decks/pack, 4 packs/case
Expand
View Pricing
CLS4129
Corning
®
DeckWorks
™
standard pipet tips,
50 - 1250μL extended length pipet tips, graduated, reload system, natural, non-sterile, polypropylene, 96 tips/deck, 8 decks/pack, 4 packs/case
Expand
View Pricing
CLS4125
Corning
®
DeckWorks
™
standard pipet tips,
0.1 - 10μL pipet tips, graduated, reload system, natural, non-sterile, polypropylene, 96 tips/deck, 10 decks/pack, 4 packs/case
Expand
View Pricing
CLS4127
Corning
®
DeckWorks
™
standard pipet tips,
1 - 200μL pipet tips, graduated, reload system, yellow, non-sterile, polypropylene, 96 tips/deck, 10 decks/pack, 4 packs/case
Expand
View Pricing
CLS4110
Corning
®
DeckWorks
™
standard pipet tips,
0.1 - 10μL extended length pipet tips, graduated, natural, non-sterile, polypropylene, 1000 tips/bag, 10,000 tips/case
Expand
View Pricing
CLS4123
Corning
®
DeckWorks
™
standard pipet tips,
1 - 300μL extended length pipet tips, graduated, hinged racks, natural, sterile, polypropylene, 96 tips/rack, 10 racks/pack, 4 packs/case
Expand
View Pricing
CLS4121
Corning
®
DeckWorks
™
standard pipet tips,
1 - 200μL pipet tips, graduated, hinged racks, natural, sterile, polypropylene, 96 tips/rack, 10 racks/pack, 4 packs/case
Expand
View Pricing
CLS4124
Corning
®
DeckWorks
™
standard pipet tips,
50 - 1250μL extended length pipet tips, graduated, hinged racks, natural, sterile, polypropylene, 96 tips/rack, 8 racks/pack, 4 packs/case
Expand
View Pricing
CLS4115
Corning
®
DeckWorks
™
standard pipet tips,
0.1 - 10μL extended length pipet tips, graduated, hinged racks, natural, non-sterile, polypropylene, 96 tips/rack, 10 racks/pack, 4 packs/case
Expand
View Pricing
CLS4111
Corning
®
DeckWorks
™
standard pipet tips,
1 - 200μL pipet tips, graduated, natural, non-sterile, polypropylene, 1000 tips/pack, 10,000 tips/case
Expand
View Pricing
CLS4112
Corning
®
DeckWorks
™
standard pipet tips,
1 - 200μL pipet tips, graduated, yellow, non-sterile, polypropylene, 1000 tips/pack, 10,000 tips/case
Expand
View Pricing
CLS4119
Corning
®
DeckWorks
™
standard pipet tips,
50 - 1250μL extended length pipet tips, graduated, hinged Racks, natural, non-sterile, polypropylene, 96 tips/rack, 8 racks/pack, 4 racks/case
Expand
View Pricing
CLS4120
Corning
®
DeckWorks
™
standard pipet tips,
0.1 - 10μL extended length pipet tips, graduated, hinged racks, natural, sterile, polypropylene, 96 tips/rack, 10 racks/pack, 4 packs/case
Expand
View Pricing
CLS4128
Corning
®
DeckWorks
™
standard pipet tips,
1 - 300μL extended length pipet tips, graduated, reload system, natural, non-sterile, polypropylene, 96 tips/deck, 10 decks/pack, 4 racks/case
Expand
View Pricing
CLS4118
Corning
®
DeckWorks
™
standard pipet tips,
1 - 300μL pipet tips, graduated, hinged racks, natural, non-sterile, polypropylene, 96 tips/rack, 10 racks/pack, 4 packs/case
Expand
View Pricing
CLS4116
Corning
®
DeckWorks
™
standard pipet tips,
1 - 200μL pipet tips, graduated, hinged racks, natural, non-sterile, polypropylene, 96 tips/rack, 10 racks/pack, 4 packs/case
Expand
View Pricing
CLS4117
Corning
®
DeckWorks
™
standard pipet tips,
1 - 200μL pipet tips, graduated, hinged racks, yellow, non-sterile, polypropylene, 96 tips/rack, 10 racks/pack, 4 packs/case
Expand
View Pricing
CLS4122
Corning
®
DeckWorks
™
standard pipet tips,
1 - 200μL pipet tips, graduated, hinged racks, yellow, sterile, polypropylene, 96 tips/rack, 10 racks/pack, 4 packs/case
Expand
View Pricing