Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Corning
®
cryogenic vials, external thread
Products
Product Category
vials (7)
Color
Manufacturer
Capacity (mL)
Color
Package Size
Sterility
Sterilization Method
Technique
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
CLS430659
Corning
®
cryogenic vials, external thread,
capacity 2.0 mL
Expand
View Pricing
CLS430658
Corning
®
cryogenic vials, external thread,
capacity 1.2 mL
Expand
View Pricing
CLS430663
Corning
®
cryogenic vials, external thread,
capacity 5.0 mL
Expand
View Pricing
CLS8671
Corning
®
cryogenic vials, external thread,
capacity 2.0 mL, 1D and 2D barcodes
Expand
View Pricing
CLS430661
Corning
®
cryogenic vials, external thread,
capacity 2.0 mL
Expand
View Pricing
CLS430662
Corning
®
cryogenic vials, external thread,
capacity 4.0 mL
Expand
View Pricing
CLS8676
Corning
®
cryogenic vials, external thread,
capacity 2.0 mL, 1D barecodes
Expand
View Pricing