Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Corning
®
Costar
®
Spin-X
®
centrifuge tube filters
Products
Product Category
centrifugal filters (7)
Brand
Sigma-Aldrich (2)
Manufacturer
Color
Housing Material
Membrane Material
Package Size
Pore size (µm)
Sterility
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
CLS8160
Corning
®
Costar
®
Spin-X
®
centrifuge tube filters,
cellulose acetate membrane, pore size 0.22 μm, sterile
Expand
View Pricing
CLS8161
Corning
®
Costar
®
Spin-X
®
centrifuge tube filters,
cellulose acetate membrane, pore size 0.22 μm, non-sterile
Expand
View Pricing
CLS8162
Corning
®
Costar
®
Spin-X
®
centrifuge tube filters,
cellulose acetate membrane, pore size 0.45 μm, sterile
Expand
View Pricing
CLS8169
Corning
®
Costar
®
Spin-X
®
centrifuge tube filters,
Nylon membrane, pore size 0.22 μm, non-sterile
Expand
View Pricing
CLS8163
Corning
®
Costar
®
Spin-X
®
centrifuge tube filters,
cellulose acetate membrane, pore size 0.45 μm, non-sterile
Expand
View Pricing
CLS8170
Corning
®
Costar
®
Spin-X
®
centrifuge tube filters,
Nylon membrane, pore size 0.45 μm, non-sterile
Expand
View Pricing
CLS9301
Corning
®
Costar
®
Spin-X
®
centrifuge tube filters,
without membrane, non-sterile
Expand
View Pricing