Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Corning
®
Costar
®
Not Treated Multiple Well Plates
Products
Product Category
plates (3)
Manufacturer
Binding Type
Volume (ml)
Wells
Color
Package Size
Sterility
Sterilization Method
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
CLS3738
Corning
®
Costar
®
Not Treated Multiple Well Plates,
size 24 wells, clear, polystyrene, flat bottom, case of 100 (20 Bulk Packs of 5), sterile, lid
Expand
View Pricing
CLS3736
Corning
®
Costar
®
Not Treated Multiple Well Plates,
size 6 wells, clear, polystyrene, flat bottom, case of 100 (20 Bulk Packs of 5), sterile, lid
Expand
View Pricing
CLS3737
Corning
®
Costar
®
Not Treated Multiple Well Plates,
size 12 wells, clear, polystyrene, flat bottom, case of 100 (20 Bulk Packs of 5), sterile, lid
Expand
View Pricing