Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Corning
®
CoolRack
®
Products
Product Category
racks (36)
Color
Manufacturer
Diameter (mm)
Length (cm)
Technique
Filter Products
Showing 1-20 of 36
Page 1 of 2
Page 1 of 2
Compare
Product Number
Description
Pricing
CLS432045
Corning
®
CoolRack
®
,
M96, capacity 96 x 1.5 or 2 mL microfuge tubes with A-H and 1-12 row and column Indexing, gray
Expand
View Pricing
CLS432053
Corning
®
CoolRack
®
,
XT PCR96, capacity 12 strip wells
Expand
View Pricing
CLS432041
Corning
®
CoolRack
®
,
M30, capacity 30 x 1.5 or 2 mL microfuge tubes, gray
Expand
View Pricing
CLS432051
Corning
®
CoolRack
®
,
CF45, capacity 45 cryovial or FACS tubes
Expand
View Pricing
CLS432054
Corning
®
CoolRack
®
,
XT-M-PCR, capacity 12 x 1.5 or 2 mL microfuge tubes and 6 strip wells
Expand
View Pricing
CLS432061
Corning
®
CoolRack
®
,
15 ml, capacity 9 x 15 mL conical centrifuge tubes
Expand
View Pricing
CLS432060
Corning
®
CoolRack
®
,
XT 5ml, capacity 12 x 5 mL microfuge tubes
Expand
View Pricing
CLS432062
Corning
®
CoolRack
®
,
50 ml, capacity 4 x 50 mL conical centrifuge tubes
Expand
View Pricing
CLS432034
Corning
®
CoolRack
®
,
M6, capacity 6 x 1.5 or 2 mL microfuge tubes, gray
Expand
View Pricing
CLS432049
Corning
®
CoolRack
®
,
CF15, capacity 15 cryovial or FACS tubes
Expand
View Pricing
CLS432042
Corning
®
CoolRack
®
,
M30, capacity 30 x 1.5 or 2 mL microfuge tubes, green
Expand
View Pricing
CLS432040
Corning
®
CoolRack
®
,
XTM24, capacity 24 x 1.5 or 2mL microfuge tubes
Expand
View Pricing
CLS432055
Corning
®
CoolRack
®
,
XT PCR384
Expand
View Pricing
CLS432037
Corning
®
CoolRack
®
,
M15, capacity 15 x 1.5 or 2 mL microfuge tubes, gray
Expand
View Pricing
CLS432052
Corning
®
CoolRack
®
,
CFT30, capacity 30 cryovial or FACS tubes, with "gripping" wells for one-hand Vial opening/closing
Expand
View Pricing
CLS432035
Corning
®
CoolRack
®
,
M6, capacity 6 x 1.5 or 2 mL microfuge tubes, green
Expand
View Pricing
CLS432063
Corning
®
CoolRack
®
,
250 ml, capacity 1 x 250 mL conical centrifuge tubes
Expand
View Pricing
CLS432038
Corning
®
CoolRack
®
,
M15, capacity 15 x 1.5 or 2 mL microfuge tubes, green
Expand
View Pricing
CLS432044
Corning
®
CoolRack
®
,
M90, capacity 90 x 1.5 or 2 mL microfuge tubes, gray
Expand
View Pricing
CLS432043
Corning
®
CoolRack
®
,
M30, capacity 30 x 1.5 or 2 mL microfuge tubes, orange
Expand
View Pricing
Page 1 of 2
Page 1 of 2