Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Corning
®
96 Well TC-Treated Microplates
Products
Product Category
plates (8)
Color
Manufacturer
Binding Type
Diameter (mm)
Volume (µL)
Wells
Color
Package Size
Sterility
Sterilization Method
Filter Products
Showing 1-8 of 8
Compare
Product Number
Description
Pricing
CLS3596
Corning
®
96 Well TC-Treated Microplates,
size 96 wells, polystyrene, flat bottom, case of 50 (individually wrapped), sterile, lid
Expand
View Pricing
CLS3799
Corning
®
96 Well TC-Treated Microplates,
size 96 wells, clear, polystyrene, round bottom, case of 50 (individually wrapped), sterile, lid
Expand
View Pricing
CLS3595
Corning
®
96 Well TC-Treated Microplates,
size 96 wells, polystyrene plate, flat bottom, case of 50 (individually wrapped), sterile, lid (low evaporation)
Expand
View Pricing
CLS3599
Corning
®
96 Well TC-Treated Microplates,
size 96 wells, polystyrene, flat bottom, case of 100 (individually wrapped), sterile, lid
Expand
View Pricing
CLS3997
Corning
®
96 Well TC-Treated Microplates,
size 96 wells, clear, polystyrene, flat bottom, case of 50 (10 Bulk Packs of 5), sterile, lid
Expand
View Pricing
CLS3894
Corning
®
96 Well TC-Treated Microplates,
size 96 wells, clear, polystyrene, V-bottom, case of 50 (individually wrapped), sterile, lid
Expand
View Pricing
CLS3598
Corning
®
96 Well TC-Treated Microplates,
size 96 wells, polystyrene, flat bottom, case of 100 (20 Bulk Packs of 5), sterile, lid
Expand
View Pricing
CLS3585
Corning
®
96 Well TC-Treated Microplates,
size 96 wells, polystyrene, flat bottom, case of 50 (10 Bulk Packs of 5), sterile, lid (low evaporation)
Expand
View Pricing