Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Corning
®
96 well NBS
™
Microplate
Products
Product Category
plates (8)
Color
Manufacturer
Binding Type
Volume (µL)
Wells
Color
Package Size
Sterility
Filter Products
Showing 1-8 of 8
Compare
Product Number
Description
Pricing
CLS3991
Corning
®
96 well NBS
™
Microplate,
flat bottom, black polystyrene, bag of 5 ×, non-sterile, lid: no
Expand
View Pricing
CLS3605
Corning
®
96 well NBS
™
Microplate,
round bottom, white polystyrene, bag of 25 ×, non-sterile, lid: no
Expand
View Pricing
CLS3641
Corning
®
96 well NBS
™
Microplate,
flat bottom clear, polystyrene, bag of 25 ×, non-sterile, lid: no
Expand
View Pricing
CLS3651
Corning
®
96 well NBS
™
Microplate,
Expand
View Pricing
CLS3650
Corning
®
96 well NBS
™
Microplate,
flat bottom, black polystyrene, bag of 25 ×, non-sterile, lid: no
Expand
View Pricing
CLS3600
Corning
®
96 well NBS
™
Microplate,
flat bottom, white polystyrene, bag of 25 ×, non-sterile, lid: no
Expand
View Pricing
CLS3990
Corning
®
96 well NBS
™
Microplate,
flat bottom, white polystyrene, bag of 5 ×, non-sterile, lid: no
Expand
View Pricing
CLS3604
Corning
®
96 well NBS
™
Microplate,
flat bottom clear, white polystyrene, bag of 25 ×, non-sterile, lid: no
Expand
View Pricing