Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Corning
®
50 mL centrifuge tubes
Products
Product Category
tubes (7)
Color
Manufacturer
Agency
Capacity (mL)
Diameter (mm)
OD (mm)
Color
Package Size
Special Grade
Natural (6)
Sterility
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
CLS430829
Corning
®
50 mL centrifuge tubes,
50 mL centrifuge tubes, polypropylene, conical bottom w/ CentriStar cap, bulk packed, sterile, natural, 500/cs
Expand
View Pricing
CLS430828
Corning
®
50 mL centrifuge tubes,
50 mL centrifuge tubes, polypropylene, conical bottom w/ CentriStar cap, rack packed, sterile, natural, 25/rack, 500/cs
Expand
View Pricing
CLS430290
Corning
®
50 mL centrifuge tubes,
50 mL centrifuge tubes, polypropylene, conical bottom w/ plug seal cap, rack packed, sterile, natural, 25/rack, 500/cs
Expand
View Pricing
CLS4558
Corning
®
50 mL centrifuge tubes,
50 mL centrifuge tubes, polypropylene, conical bottom w/ CentriStar cap, universal rack packed, sterile, natural, 25/universal rack, 300/cs
Expand
View Pricing
CLS430291
Corning
®
50 mL centrifuge tubes,
50 mL centrifuge tubes, polypropylene, conical bottom w/ plug seal cap, bulk packed, sterile, natural, 500/cs
Expand
View Pricing
CLS431472
Corning
®
50 mL centrifuge tubes,
50 mL centrifuge tubes, polypropylene, conical tube w/ no cap, bulk packed, sterile, natural, 500/cs
Expand
View Pricing
CLS430304
Corning
®
50 mL centrifuge tubes,
conical clear PET bottom, sterile, cap (plug seal), self-standing: no (rack packed), case of 500
Expand
View Pricing