Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Corning
®
384 well microplate, low volume
Products
Product Category
plates (17)
Color
Manufacturer
Binding Type
Volume (µL)
Wells
Color
Package Size
Sterility
Filter Products
Showing 1-17 of 17
Compare
Product Number
Description
Pricing
CLS4512
Corning
®
384 well microplate, low volume,
white polystyrene, round bottom, non-sterile, lid: no, pack of 10
Expand
View Pricing
CLS4514
Corning
®
384 well microplate, low volume,
surface treatment, Non-binding Surface, black polystyrene, U-bottom, non-sterile, lid: no, pack of 10
Expand
View Pricing
CLS4513
Corning
®
384 well microplate, low volume,
surface treatment, Non-binding Surface, white polystyrene, U-bottom, non-sterile, lid: no, U-bottom (white), pack of 10
Expand
View Pricing
CLS4511
Corning
®
384 well microplate, low volume,
black polystyrene, round bottom, non-sterile, lid: no, non-treated, pack of 10
Expand
View Pricing
CLS3544
Corning
®
384 well microplate, low volume,
NBS
™
(non-binding surface), black polystyrene, flat bottom, non-sterile, lid: no, pack of 10
Expand
View Pricing
CLS3540
Corning
®
384 well microplate, low volume,
black polystyrene, round bottom, non-sterile, lid: no, pack of 10
Expand
View Pricing
CLS3821
Corning
®
384 well microplate, low volume,
black polystyrene, flat bottom, non-sterile, lid: no, pack of 10
Expand
View Pricing
CLS3824
Corning
®
384 well microplate, low volume,
NBS
™
(non-binding surface), white polystyrene, flat bottom, non-sterile, lid: no, pack of 10
Expand
View Pricing
CLS3570BC
Corning
®
384 well microplate, low volume,
Tissue Culture (TC)-treated surface, white polystyrene, flat bottom white, sterile, with generic barcode, lid, pack of 10
Expand
View Pricing
CLS3821BC
Corning
®
384 well microplate, low volume,
black polystyrene, flat bottom, non-sterile, with generic barcode, lid: no, pack of 10
Expand
View Pricing
CLS3542
Corning
®
384 well microplate, low volume,
black polystyrene, flat bottom, sterile, lid, pack of 50
Expand
View Pricing
CLS3826
Corning
®
384 well microplate, low volume,
Tissue Culture (TC)-treated surface, white polystyrene, flat bottom, sterile, lid, pack of 10
Expand
View Pricing
CLS3824BC
Corning
®
384 well microplate, low volume,
white polystyrene, flat bottom, non-sterile, with generic barcode, lid: no, pack of 10
Expand
View Pricing
CLS3822
Corning
®
384 well microplate, low volume,
Tissue Culture (TC)-treated surface, black polystyrene, round bottom, sterile, lid, pack of 10
Expand
View Pricing
CLS3643
Corning
®
384 well microplate, low volume,
poly-D-lysine coated surface, black polystyrene, round bottom, sterile, lid, pack of 10
Expand
View Pricing
CLS4510
Corning
®
384 well microplate, low volume,
High Binding surface treatment, black polystyrene, round bottom, non-sterile, lid: no, pack of 10
Expand
View Pricing
CLS3571BC
Corning
®
384 well microplate, low volume,
Tissue Culture (TC)-treated surface, black polystyrene, flat bottom, sterile, with generic barcode, lid, pack of 10
Expand
View Pricing